siRNA Details
| Gene Symbol: Zfp148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011749.4 | NP_035879.1 | 4 | 566 | ||
| GenBank mRNA | AF316548.1 | - | 4 | 505 | ||
| AK146591.1 | - | 4 | 428 | |||
| BC026144.1 | - | 4 | 431 | 3991738 | ||
| CK634070.1 | - | 2 | 214 | |||
| U80078.1 | - | 1 | 90 | |||
| X98096.1 | - | 4 | 463 | |||

Zfp148
Genomic Map