siRNA Details
| Gene Symbol: Tulp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011657.2 | NP_035787.1 | 4 | 369 | ||
| GenBank mRNA | AB016963.1 | - | 4 | 312 | ||
| AF045582.1 | - | 4 | 322 | |||
| AK035363.1 | - | 4 | 334 | |||
| AK142527.1 | - | 4 | 359 | |||
| BC060068.1 | - | 4 | 299 | 6419643 | ||

Tulp3
Genomic Map