siRNA Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011672.4 | NP_035802.3 | 2, 3 | 256 | ||
| GenBank mRNA | AK010461.1 | - | 2, 3 | 225 | ||
| AK012543.1 | - | 204 | ||||
| AK088823.1 | - | 2, 3 | 225 | |||
| AK147138.1 | - | 2, 3 | 239 | |||
| AK169062.1 | - | 2, 3 | 238 | |||
| BC006630.1 | - | 2, 3 | 213 | 3708320 | ||
| CJ085208.1 | - | 2, 3 | 256 | |||
| U64445.1 | - | 2, 3 | 241 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053418, CK599711, BC161818, AF234601.

Ufd1l
Genomic Map