siRNA Details
| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185164.1 | NP_001172093.1 | 7 | 924 | ||
| NM_011697.3 | NP_035827.1 | 7 | 1025 | |||
| GenBank mRNA | AK148188.1 | - | 7 | 1024 | ||
| AK210361.1 | - | 1 | 90 | |||
| AK214445.1 | - | 1 | 90 | |||
| BC046303.1 | - | 7 | 902 | 6492277 | ||
| U43836.1 | - | 7 | 977 | |||
| U48800.1 | - | 7 | 779 | |||
| U52820.1 | - | 7 | 880 | |||

Vegfb
Genomic Map