siRNA Details
| Gene Symbol: Dctn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293757.1 | NP_001280686.1 | 4 | 569 | ||
| NM_001293758.1 | NP_001280687.1 | 3 | 402 | |||
| NM_001293759.1 | NP_001280688.1 | 4 | 639 | |||
| NM_011722.4 | NP_035852.1 | 3 | 456 | |||
| GenBank mRNA | AF124788.1 | - | 3 | 187 | ||
| AF190796.1 | - | 3 | 197 | |||
| AK003618.1 | - | 3 | 196 | |||
| AK088312.1 | - | 3 | 203 | |||
| BC029249.1 | - | 3 | 197 | 4486579 | ||
| BY079663.1 | - | 3 | 256 | |||
| CV560309.1 | - | 3 | 353 | |||
| DV042569.1 | - | 3 | 411 | |||
| DV062065.1 | - | 4 | 361 | |||

Dctn6
Genomic Map