siRNA Details
| Gene Symbol: Zfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011767.2 | NP_035897.2 | 5 | 645 | ||
| GenBank mRNA | AF071059.1 | - | 5 | 643 | ||
| BC038599.1 | - | 5 | 649 | |||
| BC058570.1 | - | 5 | 645 | |||
| BC094220.1 | - | 4 | 533 | |||
| BC141139.1 | - | 5 | 629 | 9055751 | ||

Zfr
Genomic Map