siRNA Details
| Gene Symbol: Uchl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011670.2 | NP_035800.2 | 4 | 396 | ||
| GenBank mRNA | AB025313.1 | - | 4 | 247 | ||
| AF172334.1 | - | 4 | 235 | |||
| AK004066.1 | - | 4 | 273 | |||
| AK013729.1 | - | 4 | 276 | |||
| AK170728.1 | - | 2 | 234 | |||
| BC039177.1 | - | 4 | 262 | 4506229 | ||
| CX241178.1 | - | 4 | 396 | |||
| JX050004.1 | - | 2 | 200 | |||

Uchl1
Genomic Map