siRNA Details
| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185164.1 | NP_001172093.1 | 7 | 994 | ||
| NM_011697.3 | NP_035827.1 | 7 | 1095 | |||
| GenBank mRNA | AK148188.1 | - | 7 | 1094 | ||
| AK210361.1 | - | 1 | 160 | |||
| AK214445.1 | - | 1 | 160 | |||
| BC046303.1 | - | 7 | 972 | 6492277 | ||
| U43836.1 | - | 7 | 1047 | |||

Vegfb
Genomic Map