siRNA Details
| Gene Symbol: Wnt8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011720.3 | NP_035850.2 | 6 | 764 | ||
| GenBank mRNA | AF130349.1 | - | 6 | 766 | ||
| AK050992.1 | - | 6 | 765 | |||
| AK169459.1 | - | 2 | 264 | |||
| BC119544.2 | - | 6 | 696 | 40056929 | ||

Wnt8b
Genomic Map