siRNA Details
| Gene Symbol: Zfp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011755.2 | NP_035885.2 | 3 | 2348 | ||
| GenBank mRNA | AK132703.1 | - | 3 | 2351 | ||
| AK159080.1 | - | 3 | 2302 | |||
| X17617.1 | - | 3 | 2299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp35| Gene Symbol: Zfp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011755.2 | NP_035885.2 | 3 | 2348 | ||
| GenBank mRNA | AK132703.1 | - | 3 | 2351 | ||
| AK159080.1 | - | 3 | 2302 | |||
| X17617.1 | - | 3 | 2299 | |||