siRNA Details
| Gene Symbol: Zfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011767.2 | NP_035897.2 | 9 | 1632 | ||
| GenBank mRNA | AF071059.1 | - | 9 | 1630 | ||
| AK034963.1 | - | 2 | 201 | |||
| BC141139.1 | - | 9 | 1616 | 9055751 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfr| Gene Symbol: Zfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011767.2 | NP_035897.2 | 9 | 1632 | ||
| GenBank mRNA | AF071059.1 | - | 9 | 1630 | ||
| AK034963.1 | - | 2 | 201 | |||
| BC141139.1 | - | 9 | 1616 | 9055751 | ||