siRNA Details
| Gene Symbol: Xcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011798.4 | NP_035928.2 | 2 | 742 | ||
| GenBank mRNA | AK040878.1 | - | 2 | 743 | ||
| AK089908.1 | - | 2 | 743 | |||
| BC141368.1 | - | 1 | 649 | 9055981 | ||
| BC141369.1 | - | 1 | 649 | 9055982 | ||

Xcr1
Genomic Map