siRNA Details
| Gene Symbol: Mab21l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011839.3 | NP_035969.2 | 1 | 708 | ||
| GenBank mRNA | AF126427.1 | - | 1 | 1049 | ||
| AF149877.1 | - | 1 | 450 | |||
| AK053645.1 | - | 1 | 709 | |||
| AK144373.1 | - | 1 | 877 | |||
| BC046628.1 | - | 1 | 674 | 6493460 | ||
| BC058730.1 | - | 1 | 814 | |||

Mab21l2
Genomic Map