siRNA Details
| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF099067.1 | - | 2 | 302 | ||
| AJ223071.1 | - | 2 | 392 | |||
| AK078809.1 | - | 2 | 414 | |||
| AK132803.1 | - | 2 | 330 | |||
| AK144130.1 | - | 2 | 276 | |||
| AK147381.1 | - | 2 | 380 | |||
| AK149584.1 | - | 2 | 330 | |||
| AK168094.1 | - | 2 | 313 | |||
| BC057939.1 | - | 2 | 304 | 5325558 | ||
| Y09234.1 | - | 2 | 392 | |||

Nek4
Genomic Map