siRNA Details
| Gene Symbol: Creg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011804.2 | NP_035934.1 | 2 | 379 | ||
| GenBank mRNA | AF084524.1 | - | 2 | 379 | ||
| AK005388.1 | - | 2 | 400 | |||
| AK010947.1 | - | 2 | 396 | |||
| AK085844.1 | - | 2 | 191 | |||
| AK144101.1 | - | 2 | 398 | |||
| AK150006.1 | - | 3 | 505 | |||
| AK190952.1 | - | 2 | 88 | |||
| AK191906.1 | - | 2 | 88 | |||
| AK193253.1 | - | 2 | 94 | |||
| AK194678.1 | - | 2 | 89 | |||
| AK203453.1 | - | 2 | 88 | |||
| AK204882.1 | - | 2 | 88 | |||
| AK219109.1 | - | 2 | 83 | |||
| BC027426.1 | - | 2 | 379 | 3670649 | ||
| Gene Symbol: Fam3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK214038.1 | - | 348 | |||

Creg1
Genomic Map