siRNA Details
| Gene Symbol: Gpr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011823.4 | NP_035953.3 | 3 | 751 | ||
| GenBank mRNA | AK033675.1 | - | 3 | 733 | ||
| AK039041.1 | - | 3 | 682 | |||
| AK039528.1 | - | 3 | 730 | |||
| AK140507.1 | - | 3 | 663 | |||
| DQ103764.1 | - | 4 | 895 | |||
| DQ103765.1 | - | 5 | 930 | |||
| DQ103766.1 | - | 4 | 787 | |||
| DQ103767.1 | - | 3 | 752 | |||
More Information
Other Species Targeted
Set Membership: |
|

Gpr34
Genomic Map