siRNA Details
| Gene Symbol: Tenm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290702.1 | NP_001277631.1 | 1, 2 | 309 | ||
| NM_011856.4 | NP_035986.3 | 1, 2 | 309 | |||
| GenBank mRNA | AB025411.1 | - | 1, 2 | 346 | ||
| BC141361.1 | - | 1, 2 | 310 | 9055973 | ||

Tenm2
Genomic Map