siRNA Details
| Gene Symbol: Adam23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011780.2 | NP_035910.1 | 26 | 2965 | ||
| GenBank mRNA | AB009673.1 | - | 26 | 2737 | ||
| AK029301.1 | - | 24 | 2280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam23| Gene Symbol: Adam23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011780.2 | NP_035910.1 | 26 | 2965 | ||
| GenBank mRNA | AB009673.1 | - | 26 | 2737 | ||
| AK029301.1 | - | 24 | 2280 | |||