siRNA Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011790.4 | NP_035920.1 | 16 | 2117 | ||
| GenBank mRNA | AJ130975.1 | - | 15 | 1977 | ||
| AK045012.1 | - | 2100 | ||||
| AK088772.1 | - | 1 | 1843 | |||
| AK149936.1 | - | 16 | 2097 | |||
| AK167104.1 | - | 16 | 2075 | |||
| AK168055.1 | - | 16 | 2120 | |||
| BC051998.1 | - | 16 | 2117 | 6400287 | ||
| BC052422.1 | - | 16 | 2117 | 6400287 | ||

Arih2
Genomic Map