siRNA Details
| Gene Symbol: Katna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011835.2 | NP_035965.2 | 3 | 346 | ||
| GenBank mRNA | AF153197.1 | - | 3 | 324 | ||
| AK012319.1 | - | 3 | 346 | |||
| AK041900.1 | - | 3 | 341 | |||
| AK208851.1 | - | 2 | 203 | |||
| BC009136.1 | - | 3 | 260 | 3597742 | ||

Katna1
Genomic Map