siRNA Details
| Gene Symbol: Nek3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162947.1 | NP_001156419.1 | 15 | 1662 | ||
| NM_011848.4 | NP_035978.2 | 16 | 1714 | |||
| GenBank mRNA | AF093416.1 | - | 15 | 1667 | ||
| AF099066.1 | - | 16 | 1683 | |||

Nek3
Genomic Map