siRNA Details
| Gene Symbol: Ing1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA407185.1 | - | 2 | 178 | ||
| AB221554.1 | - | 2 | 161 | |||
| AF149820.1 | - | 2 | 155 | |||
| AF177755.1 | - | 2 | 143 | |||
| AF177756.1 | - | 2 | 128 | |||
| AF177757.1 | - | 2 | 1001 | |||
| AK035490.1 | - | 2 | 158 | |||
| AK076482.1 | - | 2 | 51 | |||
| AK141466.1 | - | 2 | 51 | |||
| AK145553.1 | - | 2 | 53 | |||
| AK157424.1 | - | 2 | 150 | |||
| BC016573.1 | - | 2 | 543 | |||
| BC147770.1 | - | 2 | 253 | 9087493 | ||
| BC147784.1 | - | 2 | 253 | 9087503 | ||
| BY063246.1 | - | 2 | 177 | |||
| CA320790.1 | - | 2 | 588 | |||
| CK624100.1 | - | 2 | 39 | |||
| DV655694.1 | - | 2 | 121 | |||

Ing1
Genomic Map