siRNA Details
| Gene Symbol: Grk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038018.4 | NP_001033107.1 | 2 | 259 | ||
| NM_001112711.2 | NP_001106182.1 | 2 | 259 | |||
| NM_001286063.1 | NP_001272992.1 | 2 | 239 | |||
| NM_001286064.1 | NP_001272993.1 | 2 | 259 | |||
| NM_001286065.1 | NP_001272994.1 | 2 | 259 | |||
| NM_001286066.1 | NP_001272995.1 | 2 | 259 | |||
| NM_011938.4 | NP_036068.2 | 2 | 259 | |||
| GenBank mRNA | AF040747.1 | - | 2 | 129 | ||
| AF040748.1 | - | 2 | 129 | |||
| AF040749.1 | - | 2 | 129 | |||
| AK051405.1 | - | 2 | 240 | |||
| AK146119.1 | - | 2 | 134 | |||
| BC057206.1 | - | 2 | 248 | |||
| BC075719.1 | - | 2 | 140 | 5031455 | ||
| CJ179645.1 | - | 2 | 260 | |||
| Y15797.1 | - | 2 | 81 | |||
| Y15798.1 | - | 2 | 103 | |||
| Y15799.1 | - | 2 | 86 | |||
| Y15800.1 | - | 2 | 81 | |||

Grk6
Genomic Map