siRNA Details
| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177313.1 | NP_001170784.1 | 4 | 378 | ||
| NM_011958.3 | NP_036088.3 | 4 | 378 | |||
| GenBank mRNA | AJ003140.1 | - | 4 | 339 | ||
| AK010258.1 | - | 4 | 362 | |||
| AK011550.1 | - | 4 | 344 | |||
| AK012669.1 | - | 4 | 346 | |||
| AK049534.1 | - | 4 | 346 | |||
| AK076324.1 | - | 4 | 379 | |||
| AK084326.1 | - | 4 | 347 | |||
| AK145454.1 | - | 4 | 389 | |||
| AK169291.1 | - | 4 | 362 | |||
| BC015072.1 | - | 4 | 350 | 4212266 | ||
| BC019748.1 | - | 4 | 320 | 5007346 | ||
| Y16386.1 | - | 3 | 210 | |||

Orc4
Genomic Map