siRNA Details
| Gene Symbol: Pclo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110796.2 | NP_001104266.1 | 1, 2 | 551 | ||
| NM_011995.4 | NP_036125.4 | 1, 2 | 551 | |||
| GenBank mRNA | AB083477.1 | - | 1, 2 | 476 | ||
| AB083478.1 | - | 1, 2 | 476 | |||
| Y19185.2 | - | 1, 2 | 551 | |||
| Y19186.2 | - | 1, 2 | 551 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001110797, NM_020098, AF138789, AF227534.

Pclo
Genomic Map