siRNA Details
| Gene Symbol: Tpra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011906.2 | NP_036036.2 | 8 | 968 | ||
| GenBank mRNA | AF051098.1 | - | 8 | 775 | ||
| AK089317.1 | - | 8 | 839 | |||
| AK155116.1 | - | 8 | 912 | |||
| AK164913.1 | - | 8 | 725 | |||
| AK172045.1 | - | 8 | 899 | |||
| BC010244.1 | - | 6 | 524 | |||
| BC028977.1 | - | 8 | 882 | 5363085 | ||
| BC057296.1 | - | 8 | 901 | |||

Tpra1
Genomic Map