siRNA Details
| Gene Symbol: Trex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011907.4 | NP_036037.1 | 2 | 825 | ||
| GenBank mRNA | AF151108.1 | - | 2 | 812 | ||
| BC104384.1 | - | 1 | 790 | 40050180 | ||
| BC104385.2 | - | 1 | 790 | 40050181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trex2| Gene Symbol: Trex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011907.4 | NP_036037.1 | 2 | 825 | ||
| GenBank mRNA | AF151108.1 | - | 2 | 812 | ||
| BC104384.1 | - | 1 | 790 | 40050180 | ||
| BC104385.2 | - | 1 | 790 | 40050181 | ||