siRNA Details
| Gene Symbol: Esrrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159500.1 | NP_001152972.1 | 2, 3 | 840 | ||
| NM_011934.4 | NP_036064.3 | 2, 3 | 739 | |||
| GenBank mRNA | AK032025.1 | - | 2, 3 | 547 | ||
| AK044339.1 | - | 2, 3 | 740 | |||
| AK052256.1 | - | 2, 3 | 842 | |||
| BC044858.1 | - | 1, 2 | 409 | |||
| BC132595.1 | - | 2, 3 | 704 | 40130872 | ||
| BC132597.1 | - | 2, 3 | 705 | 40130874 | ||
| S82458.1 | - | 1, 2 | 447 | |||
| X89594.1 | - | 1, 2 | 447 | |||

Esrrb
Genomic Map