siRNA Details
| Gene Symbol: Spry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF176903.1 | - | 3 | 715 | ||
| AK140405.1 | - | 3 | 727 | |||
| BC039139.1 | - | 3 | 733 | 3672353 | ||
| BC051977.1 | - | 2 | 1019 | |||
| BC053428.1 | - | 3 | 697 | 6506766 | ||
| BC069914.1 | - | 1 | 236 | 5251876 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: BC169059.

Spry1
Genomic Map