siRNA Details
| Gene Symbol: Mapk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011950.2 | NP_036080.2 | 4, 5 | 510 | ||
| GenBank mRNA | AK146993.1 | - | 4, 5 | 498 | ||
| BC001992.1 | - | 4, 5 | 479 | 3492903 | ||
| U81823.1 | - | 4, 5 | 467 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mapk13| Gene Symbol: Mapk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011950.2 | NP_036080.2 | 4, 5 | 510 | ||
| GenBank mRNA | AK146993.1 | - | 4, 5 | 498 | ||
| BC001992.1 | - | 4, 5 | 479 | 3492903 | ||
| U81823.1 | - | 4, 5 | 467 | |||