siRNA Details
| Gene Symbol: Pclo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110796.2 | NP_001104266.1 | 2 | 738 | ||
| NM_011995.4 | NP_036125.4 | 2 | 738 | |||
| GenBank mRNA | AB083477.1 | - | 2 | 663 | ||
| AB083478.1 | - | 2 | 663 | |||
| Y19185.2 | - | 2 | 738 | |||
| Y19186.2 | - | 2 | 738 | |||

Pclo
Genomic Map