siRNA Details
| Gene Symbol: Cops7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164089.1 | NP_001157561.1 | 5 | 581 | ||
| NM_012003.2 | NP_036133.1 | 5 | 566 | |||
| GenBank mRNA | AF071316.1 | - | 5 | 568 | ||
| AK010417.1 | - | 5 | 576 | |||
| AK010726.1 | - | 5 | 551 | |||
| AK013155.1 | - | 5 | 534 | |||
| BC003724.1 | - | 4 | 447 | 3593979 | ||
| CF898433.1 | - | 1 | 62 | |||

Cops7a
Genomic Map