siRNA Details
| Gene Symbol: Ctsj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012007.1 | NP_036137.1 | 4 | 440 | ||
| GenBank mRNA | AF136272.1 | - | 4 | 440 | ||
| AF158182.1 | - | 3 | 380 | |||
| AK005526.1 | - | 4 | 447 | |||
| AK131661.1 | - | 4 | 447 | |||
| BC103769.1 | - | 4 | 436 | 30787117 | ||

Ctsj
Genomic Map