siRNA Details
| Gene Symbol: Ddx3y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012008.2 | NP_036138.1 | 2 | 186 | ||
| GenBank mRNA | AJ007376.1 | - | 2 | 153 | ||
| AK154382.1 | - | 2 | 146 | |||
| BC021453.1 | - | 2 | 151 | 5043470 | ||
| BC051046.1 | - | 2 | 155 | |||
| CD552910.1 | - | 2 | 186 | |||

Ddx3y
Genomic Map