siRNA Details
| Gene Symbol: Pnck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199351.1 | NP_001186280.1 | 5 | 481 | ||
| NM_001199352.1 | NP_001186281.1 | 5 | 345 | |||
| NM_012040.3 | NP_036170.1 | 5 | 421 | |||
| GenBank mRNA | AB023027.1 | - | 4 | 291 | ||
| AF181984.1 | - | 5 | 395 | |||
| AK134665.1 | - | 5 | 352 | |||
| AK158755.1 | - | 5 | 346 | |||
| BC051996.1 | - | 5 | 328 | |||
| BC055891.1 | - | 5 | 321 | 4208888 | ||
| CX243772.1 | - | 5 | 481 | |||

Pnck
Genomic Map