siRNA Details
Gene Symbol: Omd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012050.2 | NP_036180.1 | 3 | 376 | ||
GenBank mRNA | AB007848.1 | - | 3 | 381 | ||
BC095997.1 | - | 2 | 182 | 30739852 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Omd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012050.2 | NP_036180.1 | 3 | 376 | ||
GenBank mRNA | AB007848.1 | - | 3 | 381 | ||
BC095997.1 | - | 2 | 182 | 30739852 |