siRNA Details
| Gene Symbol: Cops7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164089.1 | NP_001157561.1 | 5 | 638 | ||
| NM_012003.2 | NP_036133.1 | 5 | 623 | |||
| GenBank mRNA | AF071316.1 | - | 5 | 625 | ||
| AK010417.1 | - | 5 | 633 | |||
| AK010726.1 | - | 5 | 608 | |||
| AK019122.1 | - | 5 | 615 | |||
| BC003724.1 | - | 4 | 504 | 3593979 | ||
| CF898433.1 | - | 1 | 119 | |||

Cops7a
Genomic Map