siRNA Details
| Gene Symbol: Ppnr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | S76755.1 | - | 1 | 436 | ||
| X65588.1 | - | 1 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppnr| Gene Symbol: Ppnr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | S76755.1 | - | 1 | 436 | ||
| X65588.1 | - | 1 | 436 | |||