siRNA Details
| Gene Symbol: Pla2g2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012045.4 | NP_036175.2 | 4 | 525 | ||
| GenBank mRNA | AF166099.2 | - | 4 | 524 | ||
| AK029254.1 | - | 4 | 515 | |||
| BC125567.1 | - | 4 | 426 | 40130082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g2f| Gene Symbol: Pla2g2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012045.4 | NP_036175.2 | 4 | 525 | ||
| GenBank mRNA | AF166099.2 | - | 4 | 524 | ||
| AK029254.1 | - | 4 | 515 | |||
| BC125567.1 | - | 4 | 426 | 40130082 | ||