siRNA Details
| Gene Symbol: Aoah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012054.4 | NP_036184.1 | 5, 6 | 720 | ||
| GenBank mRNA | AK084452.1 | - | 5, 6 | 715 | ||
| AK085343.1 | - | 5, 6 | 690 | |||
| AK133301.1 | - | 5, 6 | 661 | |||
| AK170293.1 | - | 4, 5 | 256 | |||
| AK171572.1 | - | 5, 6 | 539 | |||
| BC150767.1 | - | 5, 6 | 514 | 9087678 | ||

Aoah
Genomic Map