siRNA Details
| Gene Symbol: Trpc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF139923.1 | - | 3 | 960 | ||
| AK135153.1 | - | 3 | 1149 | |||
| AK137418.1 | - | 3 | 1161 | |||
| BC117995.1 | - | 3 | 960 | 40096022 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpc7| Gene Symbol: Trpc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF139923.1 | - | 3 | 960 | ||
| AK135153.1 | - | 3 | 1149 | |||
| AK137418.1 | - | 3 | 1161 | |||
| BC117995.1 | - | 3 | 960 | 40096022 | ||