siRNA Details
| Gene Symbol: Cd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013488.2 | NP_038516.1 | 10 | 2247 | ||
| GenBank mRNA | AK153966.1 | - | 10 | 2248 | ||
| AK161775.1 | - | 10 | 2171 | |||
| BC039137.1 | - | 10 | 2226 | 3670597 | ||
| X04836.1 | - | 10 | 2248 | |||

Cd4
Genomic Map