siRNA Details
| Gene Symbol: Stom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013515.2 | NP_038543.1 | 3, 4 | 330 | ||
| GenBank mRNA | AF093620.1 | - | 3, 4 | 281 | ||
| AK077435.1 | - | 3, 4 | 298 | |||
| AK139242.1 | - | 3, 4 | 298 | |||
| AK143724.1 | - | 3, 4 | 298 | |||
| AK148975.1 | - | 3, 4 | 299 | |||
| AK149689.1 | - | 3, 4 | 298 | |||
| AK149821.1 | - | 3, 4 | 298 | |||
| AK149991.1 | - | 3, 4 | 298 | |||
| AK151129.1 | - | 3, 4 | 297 | |||
| BC003789.1 | - | 3, 4 | 293 | 3593052 | ||
| U17297.1 | - | 3, 4 | 330 | |||

Stom
Genomic Map