siRNA Details
| Gene Symbol: Ifrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013562.2 | NP_038590.2 | 2, 3 | 430 | ||
| GenBank mRNA | AK077790.1 | - | 2, 3 | 426 | ||
| AK078888.1 | - | 427 | ||||
| AK168682.1 | - | 2, 3 | 427 | |||
| BC037013.1 | - | 2, 3 | 404 | |||
| BC043723.1 | - | 2, 3 | 397 | 5369100 | ||
| X17400.1 | - | 2, 3 | 407 | |||

Ifrd1
Genomic Map