siRNA Details
Gene Symbol: Itga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC053031.1 | - | 2 | 686 | 6401146 | |
BC062205.1 | - | 1 | 461 | 4950774 | ||
BC096741.1 | - | 3 | 664 | |||
CJ089026.1 | - | 2 | 148 | |||
D13867.1 | - | 2 | 308 |
