siRNA Details
| Gene Symbol: Mb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164047.1 | NP_001157519.1 | 2 | 371 | ||
| NM_001164048.1 | NP_001157520.1 | 3 | 231 | |||
| NM_013593.3 | NP_038621.2 | 3 | 319 | |||
| GenBank mRNA | AA271284.1 | - | 3 | 231 | ||
| AK137456.1 | - | 2 | 226 | |||
| BC025172.1 | - | 3 | 331 | 3979003 | ||

Mb
Genomic Map