siRNA Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013571.2 | NP_038599.1 | 4 | 621 | ||
| GenBank mRNA | AK036566.1 | - | 4 | 677 | ||
| AK137532.1 | - | 3 | 358 | |||
| AK164833.1 | - | 4 | 742 | |||
| U43585.1 | - | 4 | 628 | |||
| X81634.1 | - | 4 | 403 | |||

Ksr1
Genomic Map