siRNA Details
| Gene Symbol: Epb4.1l4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013512.2 | NP_038540.2 | 24 | 2960 | ||
| GenBank mRNA | BC007166.1 | - | 24 | 2684 | 3490061 | |
| BC011099.1 | - | 23 | 2254 | |||
| BC013557.1 | - | 24 | 2684 | 3490326 | ||
| BC089158.1 | - | 23 | 2529 | |||
| D28818.1 | - | 23 | 2228 | |||

Epb4.1l4a
Genomic Map