siRNA Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013571.2 | NP_038599.1 | 7, 8 | 1171 | ||
| GenBank mRNA | AK036566.1 | - | 7, 8 | 1227 | ||
| AK137532.1 | - | 6, 7 | 908 | |||
| AK164833.1 | - | 7, 8 | 1293 | |||
| U43585.1 | - | 7, 8 | 1178 | |||
| X81634.1 | - | 7, 8 | 953 | |||

Ksr1
Genomic Map