siRNA Details
| Gene Symbol: Odf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113213.1 | NP_001106684.1 | 10 | 1301 | ||
| NM_001113214.1 | NP_001106685.1 | 10 | 1301 | |||
| NM_001177659.1 | NP_001171130.1 | 10 | 1304 | |||
| NM_001177661.1 | NP_001171132.1 | 9 | 1147 | |||
| NM_013615.3 | NP_038643.1 | 10 | 1274 | |||
| GenBank mRNA | AF000968.1 | - | 8 | 1121 | ||
| AF034105.1 | - | 10 | 1237 | |||
| AK143726.1 | - | 10 | 1208 | |||
| AK148523.1 | - | 10 | 1214 | |||
| AK165588.1 | - | 10 | 1205 | |||
| AK168415.1 | - | 10 | 1203 | |||
| BC057001.1 | - | 11 | 1358 | 6827260 | ||

Odf2
Genomic Map